Learn More
Medchemexpress LLC Oxindole (indolin-2-one) | 59-48-3 | MFCD00005711 | >97.0% | 133.15 g/mol | C8H7NO | 25g
SDP
Catalog No. 5000270165
Encompass_Preferred
Oxindole (Indolin-2-one) is an aromatic heterocyclic building block 2-indolinone derivatives have become lead compounds in the research of kinase inhibitors
Catalog No. 50-002-70165 Supplier Medchemexpress LLC Supplier No. HYY006125G
Oxindole (Indolin-2-one) is an aromatic heterocyclic building block 2-indolinone derivatives have become lead compounds in the research of kinase inhibitors