Learn More
Medchemexpress LLC Oxirane, 2-octyl- | 2404-44-6 | MFCD00005158 | 98.0% | 156.26 g/mol | C10H20O | 100 G
SDP
Catalog No. 5000283833
Encompass_Preferred
1,2-Epoxydecane (2-Octyloxirane) is an epoxide reagent used as a building block in organic synthesis and as a biochemical reagent for laboratory research. Its reactive oxirane ring enables ring-opening and functionalization reactions commonly employed in small-molecule synthesis and materials studies.
  • Epoxide functional group suited for nucleophilic opening reactions.
  • High purity (98.0%) appropriate for research use.
  • Chemical formula C10H20O and molecular weight 156.26 g/mol.
  • Available in laboratory-scale 100 G pack for synthesis workflows.
  • Compatible with common organic solvents and reaction conditions.
Catalog No. 50-002-83833 Supplier Medchemexpress LLC Supplier No. HYW142702100G
1,2-Epoxydecane (2-Octyloxirane) is an epoxide reagent used as a building block in organic synthesis and as a biochemical reagent for laboratory research. Its reactive oxirane ring enables ring-opening and functionalization reactions commonly employed in small-molecule synthesis and materials studies.
  • Epoxide functional group suited for nucleophilic opening reactions.
  • High purity (98.0%) appropriate for research use.
  • Chemical formula C10H20O and molecular weight 156.26 g/mol.
  • Available in laboratory-scale 100 G pack for synthesis workflows.
  • Compatible with common organic solvents and reaction conditions.