Learn More
Medchemexpress LLC Oxirane, 2-octyl- | 2404-44-6 | MFCD00005158 | ≥98.0% | 156.27 g·mol⁻¹ | C10H20O | 10 G
SDP
Catalog No. 5000282918
Encompass_Preferred
1,2-Epoxydecane (2-Octyloxirane) is an aliphatic epoxide supplied as a colorless to light-yellow liquid for research use. It has molecular formula C10H20O, molecular weight 156.27 g·mol⁻¹, CAS 2404-44-6, and is offered with ≥98.0% purity. The reagent is intended for use as a synthetic building block and for studies of epoxide reactivity; storage and handling recommendations are available in the product documentation.
  • Aliphatic epoxide useful as a synthetic building block.
  • High purity suitable for research applications.
  • Colorless to light-yellow liquid with predictable physical properties.
  • Available in small pack sizes for lab-scale work.
  • Storage stability supported by recommended temperature conditions.
Catalog No. 50-002-82918 Supplier Medchemexpress LLC Supplier No. HYW14270210G
1,2-Epoxydecane (2-Octyloxirane) is an aliphatic epoxide supplied as a colorless to light-yellow liquid for research use. It has molecular formula C10H20O, molecular weight 156.27 g·mol⁻¹, CAS 2404-44-6, and is offered with ≥98.0% purity. The reagent is intended for use as a synthetic building block and for studies of epoxide reactivity; storage and handling recommendations are available in the product documentation.
  • Aliphatic epoxide useful as a synthetic building block.
  • High purity suitable for research applications.
  • Colorless to light-yellow liquid with predictable physical properties.
  • Available in small pack sizes for lab-scale work.
  • Storage stability supported by recommended temperature conditions.