Learn More
Abcam O x iRed™ probe, pero x idase fluorogenic substrate, 5MG
Catalog No. 502215315
Encompass_Preferred
MW 257.24 Da, Purity >99%. Highly sensitive, stable peroxidase fluorogenic substrate used as a hydrogen peroxide probe.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-221-5315 Supplier Abcam Supplier No. AB1420455MG
MW 257.24 Da, Purity >99%. Highly sensitive, stable peroxidase fluorogenic substrate used as a hydrogen peroxide probe.
The product is subject to the following: Abcam Restricted Use Statement