Learn More
Cell Biolabs Inc OxiSelect™ Comet Assay Kit (3-Well Slides), 75 assays (25 slides)
Catalog No. NC0588768
Encompass_Preferred
The comet assay, or single cell gel electrophoresis assay (SCGE), is a common technique for measurement of DNA damage in individual cells. Under an electrophoretic field, damaged cellular DNA (containing fragments and strand breaks) is separated from intact DNA, yielding a classic “comet tail” shape under the microscope. The extent of DNA damage is usually visually estimated by comet tail measurement; however, image analysis software is also available for measuring various parameters. The OxiSelect™ Comet Assay is a fast and sensitive kit for the measurement of cellular DNA damage. Each kit provides sufficient reagents to perform up to 25 assays (25 slides with 3 wells each).
Catalog No. NC0588768 Supplier Cell Biolabs Inc Supplier No. STA351
Add to Cart
Add to Cart
The comet assay, or single cell gel electrophoresis assay (SCGE), is a common technique for measurement of DNA damage in individual cells. Under an electrophoretic field, damaged cellular DNA (containing fragments and strand breaks) is separated from intact DNA, yielding a classic “comet tail” shape under the microscope. The extent of DNA damage is usually visually estimated by comet tail measurement; however, image analysis software is also available for measuring various parameters. The OxiSelect™ Comet Assay is a fast and sensitive kit for the measurement of cellular DNA damage. Each kit provides sufficient reagents to perform up to 25 assays (25 slides with 3 wells each).