Learn More
Cell Biolabs Inc OxiSelect™ 96-Well Comet Assay Kit, 96 assays (1 slide)
Catalog No. NC0557588
Encompass_Preferred
The comet assay, or single cell gel electrophoresis assay (SCGE), is a common technique for measurement of DNA damage in individual cells. Under an electrophoretic field, damaged cellular DNA (containing fragments and strand breaks) is separated from intact DNA, yielding a classic “comet tail” shape under the microscope. The extent of DNA damage is usually visually estimated by comet tail measurement; however, image analysis software is also available for measuring various parameters. The OxiSelect™ Comet Assay is a fast and sensitive kit for the measurement of cellular DNA damage. Each kit provides sufficient reagents to perform up to 96 assays (one slide with 96 wells).
Catalog No. NC0557588 Supplier Cell Biolabs Inc Supplier No. STA355
Add to Cart
Add to Cart
The comet assay, or single cell gel electrophoresis assay (SCGE), is a common technique for measurement of DNA damage in individual cells. Under an electrophoretic field, damaged cellular DNA (containing fragments and strand breaks) is separated from intact DNA, yielding a classic “comet tail” shape under the microscope. The extent of DNA damage is usually visually estimated by comet tail measurement; however, image analysis software is also available for measuring various parameters. The OxiSelect™ Comet Assay is a fast and sensitive kit for the measurement of cellular DNA damage. Each kit provides sufficient reagents to perform up to 96 assays (one slide with 96 wells).