Learn More
Cell Biolabs Inc OxiSelect™ Intracellular ROS Assay Kit (Green Fluorescence), 96 assays
Catalog No. NC9878788
Encompass_Preferred
Cell Biolabs’ OxiSelect™ Intracellular ROS Assay Kit is a cell-based assay for measuring hydroxyl, peroxyl, or other reactive oxygen species activity within a cell. The assay employs the cell-permeable fluorogenic probe 2’, 7’-Dichlorodihydrofluorescin diacetate (DCFH-DA). In brief, DCFH-DA is diffused into cells and is deacetylated by cellular esterases to non-fluorescent 2’, 7’-Dichlorodihydrofluorescin (DCFH), which is rapidly oxidized to highly fluorescent 2’, 7’-Dichlorodihydrofluorescein (DCF) by ROS. The fluorescence intensity is proportional to the ROS levels within the cell cytosol. The effect of antioxidant or free radical compounds on DCF-DA can be measured against the fluorescence of the provided DCF standard. The kit has a DCF detection sensitivity limit of 10 pM. Each kit provides sufficient reagents to perform up to 96 assays, including standard curve and unknown samples.
Catalog No. NC9878788 Supplier Cell Biolabs Inc Supplier No. STA342
Cell Biolabs’ OxiSelect™ Intracellular ROS Assay Kit is a cell-based assay for measuring hydroxyl, peroxyl, or other reactive oxygen species activity within a cell. The assay employs the cell-permeable fluorogenic probe 2’, 7’-Dichlorodihydrofluorescin diacetate (DCFH-DA). In brief, DCFH-DA is diffused into cells and is deacetylated by cellular esterases to non-fluorescent 2’, 7’-Dichlorodihydrofluorescin (DCFH), which is rapidly oxidized to highly fluorescent 2’, 7’-Dichlorodihydrofluorescein (DCF) by ROS. The fluorescence intensity is proportional to the ROS levels within the cell cytosol. The effect of antioxidant or free radical compounds on DCF-DA can be measured against the fluorescence of the provided DCF standard. The kit has a DCF detection sensitivity limit of 10 pM. Each kit provides sufficient reagents to perform up to 96 assays, including standard curve and unknown samples.