Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZeptoMetrix OXItek Arylesterase/Paraoxonase (PON) Assay Kit
SDP

Catalog No. 22156781 Shop All ZeptoMetrix Corporation Products
Change view
Click to view available options
Quantity:
200 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-156-781 200 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 22-156-781 Supplier ZeptoMetrix Supplier No. 0801199
Add to Cart
Add to Cart

Includes

2 bottles of 120mL arylesterase assay buffer, 150μL of arylesterase substrate, 300μL of arylesterase/paraoxonase standard

Provides a simple, reproducible method of quantifying arylesterase activity in serum or plasma

Human serum arylesterase/paraoxonase (PON) is a polymorphic enzyme that catalyzes the hydrolysis of organophosphates such as paraoxon and aromatic carboxylic acid esters of fatty acids. Most organophosphates are neurotoxins that are widely
encountered in the diet and household.

Human serum arylesterase/paraoxonase (PON) has been shown to catalyze nearly all arylesterase and paraoxonase activities. It is a glycoprotein containing 337 amino acid residues with a molecular weight of approximately 43KD and a serum concentration of about 50mg/L.

Specifications

Certifications/Compliance ISO, 13485
Concentration 50 mg/L
Content And Storage -70°C
Form Liquid
Media Type Vial
Product Type OXItek Assay Kit
Formulation 20mM Tris/HCl Buffer, pH 8, containing 1mM CaCl2 and 4mM Phenyl Acetate as the Substrate
For Use With (Application) For quantifying Arylesterase/Paraoxonase activity in Serum or Plasma
Includes Arylesterase assay Buffer, Arylesterase Substrate, Arylesterase/Paraoxonase Standard
Quantity 200 Tests

Research purposes only. Not for in vitro diagnostic use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.