Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZeptoMetrix OXItek Thiobarbituric Acid Reactive Substances (TBARS) Assay Kit
SDP

Catalog No. 22156701 Shop All ZeptoMetrix Corporation Products
Change view
Click to view available options
Quantity:
160 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-156-701 160 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 22-156-701 Supplier ZeptoMetrix Supplier No. 0801192
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Includes

4 vials of thiobarbituric acid, 4 vials of 50mL TBARS diluent 1, 4 vials of 50mL TBARS diluent 2, 30mL of SDS solution, 20mL of MDA standard, 100mL of MDA diluent, Marbles

For screening and monitoring lipid peroxidation

  • Provides standardized, reproducible assay with consistent results
  • Rapid, easy–to–use procedure
  • Can be used with many types of samples including drugs, food products and human and animal biological tissues
  • Measures anti-oxidant activity of several compounds
  • Quality controlled reagents

Specifications

Certifications/Compliance ISO, 13485
Content And Storage 2°C to 8°C
Form Liquid
Media Type Vial
Product Type OXItek Assay Kit
For Use With (Application) Screening and monitoring Lipid peroxidation, a major indicator of oxidative stress
Includes Thiobarbituric Acid (4 vials/kit), TBARS Diluent 1 (4 x 50mL/kit), TBARS Diluent 2 (4 x 50mL/kit),SDS solution (30mL/kit), MDA Standard (20mL/kit), MDA Diluent (100mL/kit), Marbles
Quantity 160 Tests

Research purposes only. Not for in vitro diagnostic use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.