Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diversey™ Oxivir™ 1 RTU

Catalog No. 17000059
Change view
Click to view available options
Quantity:
12/Cs.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
17-000-059 12/Cs.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 17-000-059 Supplier Diversey™ Supplier No. 100850916
Add to Cart
Add to Cart

Next generation RTU disinfectant wipes

  • For operating rooms, long-term care facilities, medical facilities, animal treatment facilities, cruise ships, hotels/motels, public buildings, public restrooms, recreational facilities, retail and schools
  • Multi-surface cleaner/disinfectant
  • Environmentally responsible with no VOCs
  • No phosphates, quats or phenols
  • Breaks down to water and oxygen after use
  • AHP™ based

Specifications

Product Type Disinfectant Cleaner
Sterility Non-sterile
Appearance Colorless
Certifications/Compliance EPA, FDA
Container Type Spray Bottle
Filtered Unfiltered
Flammable No
For Use With (Application) Disinfecting in public/medical facilities
Quantity 12/Cs.
Volume (English) 32 oz.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.