Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diversey™ Oxivir™ 1 Wipes

Catalog No. 17000058
Change view
Click to view available options
Packaging Quantity:
160 Wipes/Bucket
160 Wipes/Pk., 4 Pk./Cs.
Container Type:
Bag
Bucket
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Packaging Quantity Container Type
17-000-058 160 Wipes/Bucket Bucket
17-000-044 160 Wipes/Pk., 4 Pk./Cs. Bag
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 17-000-058 Supplier Diversey™ Supplier No. 100850924
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Next generation ready-to-use, pre-wetted, one-step disinfectant cleaner wipe, based on proprietary AHP™ technology. Oxivir™ 1 Wipes are formulated to deliver broad spectrum disinfection in ONE minute or less, with 75 total claims.

  • Multi-surface cleaner/disinfectant wipes
  • Broad spectrum, multi-surface cleaner/disinfectant and sanitizer
  • Harnesses the cleaning power of hydrogen peroxide
  • Specially designed to penetrate the toughest soils
  • Fast, Effective, Responsible and Sustainable formulation
  • Environmentally-responsible with no VOCs, no added fragrance, no NPEs
  • Fast-acting, convenient formula, suitable for daily use, and contact time of 1 minute or less
  • Formulated with patented AHP™ technology containing no phosphates, quats or phenols
  • The active ingredient breaks down to water and oxygen after use

Specifications

Product Type Disinfectant Cleaner Wipes
Color White
Container Type Bucket
For Use With (Application) Cleaning surfaces
Packaging Quantity 160 Wipes/Bucket
Sterility Non-sterile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.