Learn More
MSC Solenis Wipes, 11 x 12 in, 160 Count, Unscented
Catalog No. 502839476
Encompass
These ready-to-use disinfectant cleaner wipes, powered by proprietary hydrogen peroxide (AHP) technology, provide fast, effective cleaning and disinfection. They disinfect bacteria and viruses in 60 seconds, acting as a virucide, bactericide, tuberculocide, fungicide, and non-food contact sanitizer. Effective against MRSA and Norovirus, these wipes also meet bloodborne pathogen standards and contain no added fragrance.
  • One-step cleaner disinfectant with AHP technology.
  • Effective against TB, MRSA, Norovirus, VRE, HBV, HCV, and HIV.
  • Kills most pathogens in 1-5 minutes.
  • No VOCs, added fragrance, or NPEs; active ingredient breaks down to oxygen and water.
  • Compatible with most hard, non-porous surfaces; non-irritating to eyes and skin.
  • Pre-wetted wipes for convenient cleaning and disinfection.
Catalog No. 50-283-9476 Supplier MSC Supplier No. 54426903
These ready-to-use disinfectant cleaner wipes, powered by proprietary hydrogen peroxide (AHP) technology, provide fast, effective cleaning and disinfection. They disinfect bacteria and viruses in 60 seconds, acting as a virucide, bactericide, tuberculocide, fungicide, and non-food contact sanitizer. Effective against MRSA and Norovirus, these wipes also meet bloodborne pathogen standards and contain no added fragrance.
  • One-step cleaner disinfectant with AHP technology.
  • Effective against TB, MRSA, Norovirus, VRE, HBV, HCV, and HIV.
  • Kills most pathogens in 1-5 minutes.
  • No VOCs, added fragrance, or NPEs; active ingredient breaks down to oxygen and water.
  • Compatible with most hard, non-porous surfaces; non-irritating to eyes and skin.
  • Pre-wetted wipes for convenient cleaning and disinfection.