Learn More
Kalen Biomedical LLC OXLDL 1MG
Catalog No. NC9835065
Encompass_Preferred
Human Medium Oxidized Low Density Lipoprotein is oxidized with copper sulfate. The product is provided in PBS with EDTA as a preservative. The protein concentration is 1 mg/mL and the endotoxin level is less than 0.5 EU /mg of protein.
Catalog No. NC9835065 Supplier Kalen Biomedical LLC Supplier No. 7702027
Human Medium Oxidized Low Density Lipoprotein is oxidized with copper sulfate. The product is provided in PBS with EDTA as a preservative. The protein concentration is 1 mg/mL and the endotoxin level is less than 0.5 EU /mg of protein.