Learn More
TARGETMOL CHEMICALS INC OXOADIPIC ACID 10MG
SDP
Catalog No. 502579847
Encompass_Preferred
Also available in 1 mg 2 mg 5 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Oxoadipic acid (2-Oxoadipic acid) is a key metabolite of the essential amino acids tryptophan and lysine.Important metabolite between the TCA cycle and lysine biosynthesis. Of interest for research on mitochondrial metabolite transporters. purity: 96%
Catalog No. 50-257-9847 Supplier TARGETMOL CHEMICALS INC Supplier No. T492110MG
Also available in 1 mg 2 mg 5 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Oxoadipic acid (2-Oxoadipic acid) is a key metabolite of the essential amino acids tryptophan and lysine.Important metabolite between the TCA cycle and lysine biosynthesis. Of interest for research on mitochondrial metabolite transporters. purity: 96%