Learn More
Medchemexpress LLC Oxoadipic acid | 3184-35-8 | 99.86% | 160.12 | 50 MG
SDP
Catalog No. 5000437235
Encompass_Preferred
Oxoadipic acid (Standard) is an analytical standard intended for research and analytical applications. It is a key metabolite of the essential amino acids tryptophan and lysine.
  • Analytical standard
  • Intended for research and analytical applications
  • Key metabolite of tryptophan and lysine
Catalog No. 50-004-37235 Supplier Medchemexpress LLC Supplier No. HY113227R50MG
Oxoadipic acid (Standard) is an analytical standard intended for research and analytical applications. It is a key metabolite of the essential amino acids tryptophan and lysine.
  • Analytical standard
  • Intended for research and analytical applications
  • Key metabolite of tryptophan and lysine