Learn More
Medchemexpress LLC Oxoadipic acid | 3184-35-8 | 99.9% | 160.12 | 10 MG
SDP
Catalog No. 5000435683
Encompass_Preferred
Oxoadipic acid (Standard) is the analytical standard of Oxoadipic acid. This product is intended for research and analytical applications. Oxoadipic acid is a key metabolite of the essential amino acids tryptophan and lysine.
  • Analytical standard for Oxoadipic acid
  • Intended for research and analytical applications
  • Key metabolite of tryptophan and lysine
Catalog No. 50-004-35683 Supplier Medchemexpress LLC Supplier No. HY113227R10MG
Oxoadipic acid (Standard) is the analytical standard of Oxoadipic acid. This product is intended for research and analytical applications. Oxoadipic acid is a key metabolite of the essential amino acids tryptophan and lysine.
  • Analytical standard for Oxoadipic acid
  • Intended for research and analytical applications
  • Key metabolite of tryptophan and lysine