Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Oxoadipic acid | 3184-35-8 | 99.9% | 160.12 | 50 MG
SDP

Catalog No. 5000436582
Encompass_Preferred

Oxoadipic acid is a key metabolite of the essential amino acids tryptophan and lysine. This product is suitable for various laboratory applications.

  • Endogenous metabolite
  • White to light yellow solid appearance
  • Molecular formula C6H8O5
  • Soluble in H2O and DMSO
  • Store powder at -20°C for 3 years, 4°C for 2 years
  • Store in solvent at -80°C for 6 months, -20°C for 1 month

Catalog No. 50-004-36582 Supplier Medchemexpress LLC Supplier No. HY11322750MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Oxoadipic acid is a key metabolite of the essential amino acids tryptophan and lysine. This product is suitable for various laboratory applications.

  • Endogenous metabolite
  • White to light yellow solid appearance
  • Molecular formula C6H8O5
  • Soluble in H2O and DMSO
  • Store powder at -20°C for 3 years, 4°C for 2 years
  • Store in solvent at -80°C for 6 months, -20°C for 1 month

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.