Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Oxoadipic acid (standard) | 3184-35-8 | MFCD00010004 | 99.9% | 160.12 | C6H8O5 | 5 MG
SDP

Catalog No. 5000263929
Encompass_Preferred

Oxoadipic acid is an endogenous metabolite provided as an analytical standard for research and analytical applications. It has molecular formula C6H8O5 and molecular weight 160.12, and is supplied at high purity for use as a reference in assays and method development. Small pack sizes facilitate preparation of stock solutions for analytical workflows.

  • High purity (99.9%) suitable for analytical reference use.
  • Molecular formula C6H8O5; molecular weight 160.12.
  • Provided as an analytical standard for assay and method development.
  • Available in small pack sizes for convenient standard preparation.
  • Soluble in water and DMSO, enabling easy stock solution preparation.

Catalog No. 50-002-63929 Supplier Medchemexpress LLC Supplier No. HY113227R5MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Oxoadipic acid is an endogenous metabolite provided as an analytical standard for research and analytical applications. It has molecular formula C6H8O5 and molecular weight 160.12, and is supplied at high purity for use as a reference in assays and method development. Small pack sizes facilitate preparation of stock solutions for analytical workflows.

  • High purity (99.9%) suitable for analytical reference use.
  • Molecular formula C6H8O5; molecular weight 160.12.
  • Provided as an analytical standard for assay and method development.
  • Available in small pack sizes for convenient standard preparation.
  • Soluble in water and DMSO, enabling easy stock solution preparation.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.