Learn More
AdipoGen Oxocyclopentyloxy-benzop-2-one
SDP
Supplier:  AdipoGen CDXO0047M010
Encompass_Preferred
Chemical. CAS 877774-65-7. Formula C14H12O4. MW 244.24. Synthetic Fluorogenic substrates for detecting Baeyer-Villiger monooxygenase.
Catalog No. 50-256-0329