Learn More
Thermo Scientific™ Oxoid An-Ident Discs
Catalog No. R65006 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
50 Disks
Catalog No. Quantity
R65006 50 Disks
1 options
This item is not returnable. View return policy
Catalog No. R65006 Supplier Thermo Scientific™ Supplier No. DD0006A
Add to Cart
Add to Cart
This item is not returnable. View return policy
Includes
  • Erythromycin, 60μg
  • Rifampin, 15μg
  • Colistin, 10μg
  • Penicillin, 2U
  • Kanamycin, 1000μg
  • Vancomycin, 5μg
Presumptive identification for Gram-negative anaerobic organisms
Order Info
Supplied 50 disks per cartridge; 6 cartridges/set.
TRUSTED_SUSTAINABILITY
Specifications
Detectable Analytes Gram-negative anaerobic organisms
Type Oxoid An-Ident Discs
Includes Erythromycin 60μg, Rifampin 15μg, Colistin 10μg, Penicillin 2U, Kanamycin 1000μg, Vancomycin 5μg
Quantity 50 Disks