Learn More
Description
Cultivate anaerobic microorganisms, particularly Bacteroides spp. and other fastidious anaerobes with this medium.
Cultivate anaerobic microorganisms, particularly Bacteroides spp. and other fastidious anaerobes with Thermo Scientific™ Oxoid™ Anaerobe Basal Agar (Dehydrated). Anaerobe Basal Broth is formulated from a range of nutrients which have been selected to optimize the recovery and growth of the majority of anaerobic organisms of clinical importance.
Anaerobe Basal Agar contains peptones, carefully selected to support good growth of anaerobic bacteria and yeast extract as a vitamin source. Starch is present to absorb any toxic metabolites1. Sufficient arginine is added to ensure the growth of Eubacterium lentum2, while haemin and vitamin K are growth factors required by many Bacteroides species3. Haemin is also required by Porphyromonas species. Sodium succinate improves the growth of Prevotella melaninogenica and Bacteroides species4. Sodium pyruvate is added as an energy source for asaccharolytic cocci such as Veillonella. It also acts similarly to catalase and degrades traces of hydrogen peroxide, which may be produced by the action of molecular oxygen on media components5. L-cysteine hydrochloride and dithiothreitol are reducing agents, and cysteine has also been shown to stimulate the growth of some anaerobes6.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand
Specifications
Specifications
| Description | Anaerobe Basal Broth |
| Form | Powder |
| Product Type | Microbiology Media |
| For Use With (Application) | Healthcare |
| Quantity | 500 g |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.