Learn More
Thermo Scientific™ Oxoid™ AnaeroGen™ W-Zip Compact Gas Generator System
GSA_VA
Catalog No. 23313398 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
10 Each/Pk.
Catalog No. Quantity
23-313-398 10 Each/Pk.
1 options
This item is not returnable. View return policy
Catalog No. 23-313-398 Supplier Thermo Scientific™ Supplier No. AN0010W
Add to Cart
Add to Cart
This item is not returnable. View return policy
Oxoid AnaeroGen W-Zip Compact is a simple anaerobic gas generator system for the anaerobic incubation of up to four petri dishes or an identification panel.
Oxoid AnaeroGen W-Zip Compact is a simple anaerobic gas generator system for the anaerobic incubation of up to four petri dishes or an identification panel.
  • Use 1 sachet / Compact Plastic Pouch
Includes: 10 sachets and 10 W-Zip pouches.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
TRUSTED_SUSTAINABILITY
Specifications
Type AnaeroGen W-Zip Compact
Format Compact Gas Generator Kit
Includes 10 sachets and 10 W-Zip pouches
Product Line Thermo Scientific Oxoid
Quantity 10 Each/Pk.