Description
Oxoid AnaeroJar O-Ring is for use with AnaeroJar 2.5L, Part No. AG0025A.
Specifications
Specifications
| Type | AnaeroJar O-ring |
| Product Line | Thermo Scientific Oxoid |
| Quantity | 5/Pk. |