Learn More
Thermo Scientific™ Oxoid™ Antimicrobial Susceptibility Individual disc Dispenser
Shop All Thermo Scientific ProductsDescription
Accurately place antimicrobial susceptibility discs onto your media of choice when carrying out manual Antimicrobial Susceptibility Testing methods using a self-tamping Thermo Scientific™ Oxoid™ Antimicrobial Susceptibility Individual disc Dispenser.
disc diffusion not only categorizes resistant, intermediate and sensitive organisms through quantitative results but also provides a visual indication of the following:
- Inoculum level
- Presence of contamination
- Resistant mutants
- Beta-lactamase activity
- Antagonism and synergy between antibiotics
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
Specifications
Specifications
| Certifications/Compliance | For use only with Oxoid Antimicrobial Susceptibility Discs |
| Description | Oxoid Antimicrobial Susceptibility Individual Disc Dispenser |
| Product Type | Individual Disc Dispenser |
| Quantity | 6 Ejectors/Pk. |