Learn More
Thermo Scientific™ Oxoid™ Azithromycin Antimicrobial Susceptibility discs
Catalog No. CT0906B Shop All Thermo Scientific Products
Change view
Click to view available options
Concentration:
15 μg
Catalog No. Concentration
CT0906B 15 μg
1 options
This item is not returnable. View return policy
Catalog No. CT0906B Supplier Thermo Scientific™ Supplier No. CT0906B
Add to Cart
Add to Cart
This item is not returnable. View return policy
Determine the susceptibility of microorganisms using manual AST methods and a full range of high-quality, easy-to-use antimicrobial susceptibility testing discs.
Manually determine the antibiotic susceptibility of microorganisms using Antimicrobial Susceptibility Testing (AST) methods in conjunction with reliable, easy-to-use Thermo Scientific™ Oxoid™ Azithromycin Antimicrobial Susceptibility discs.
Ease-of-use is provided by shatterproof spring-loaded cartridges, easy-to-read labeling and end-of-cartridge indicators.
disc diffusion not only categorizes resistant, intermediate and sensitive organisms through quantitative results but also provides a visual indication of the following:
  • Inoculum level
  • Presence of contamination
  • Resistant mutants
  • Beta-lactamase activity
  • Antagonism and synergy between antibiotics
Each cartridge is individually sealed with a desiccant to maintain a moisture level of <2%, ensuring long term stability.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
TRUSTED_SUSTAINABILITY
Specifications
Antimicrobial Agents Azithromycin
Certifications/Compliance Complies with CLSI, DIN, BSAC, EUCAST, SFM guidelines
Concentration 15 μg
Description Oxoid Azithromycin Antimicrobial Susceptibility Discs
Antibiotic Code AZM
Product Type Antimicrobial Susceptibility Discs
Test Type Antimicrobial Susceptibility
No. per Pack 5 Cartridges/Pk.
Packaging Type 50 Discs/Cartridge
Quantity 50 Discs/Cartridge