Learn More
Thermo Scientific™ Oxoid™ Basic Fuchsin Indicator
Catalog No. OXBR0050A Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
10 g
Catalog No. Quantity
OXBR0050A 10 g
1 options
This item is not returnable. View return policy
Catalog No. OXBR0050A Supplier Thermo Scientific™ Supplier No. BR0050A
Add to Cart
Add to Cart
This item is not returnable. View return policy
Oxoid Basic Fuchsin Indicator is a dye for use with Endo Agar Base, Part No. CM0479B and m Endo (LES) Agar Base, Part No. MM0551B.
Oxoid Basic Fuchsin Indicator is a dye for use with Endo Agar Base, Part No. CM0479B and m Endo (LES) Agar Base, Part No. MM0551B.
TRUSTED_SUSTAINABILITY
Specifications
Description Basic Fuchsin Indicator
Form Powder
Product Type Indicator Dye
Quantity 10 g