Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Oxoid™ Blaser-Wang Campylobacter Selective Supplement

Catalog No. OXSR098E Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
10/Pk.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
OXSR098E 10/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. OXSR098E Supplier Thermo Scientific™ Supplier No. SR0098E
Add to Cart
Add to Cart
This item is not returnable. View return policy

Oxoid Campylobacter Selective Supplement, Blaser-Wang isolatee Campylobacter and inhibit Candida albicans.

Oxoid Campylobacter Selective Supplement, Blaser-Wang is used to prepare of Blaser-Wang Selective Medium for the isolation of Campylobacter while inhibiting growth of Candida albicans.

  • Add to Columbia Blood Agar Base, Part No. CM0271B
  • Each vial supplements 500mL of medium
TRUSTED_SUSTAINABILITY

Specifications

Description Campylobacter Selective Supplement, Blaser-Wang
Form Freeze-dried
Product Type Selective Supplement
Quantity 10/Pk.
Yield Each vial for 500mL medium
Species Campylobacter
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.