Learn More
Description
Oxoid Brucella Selective Supplement is used for the isolation of Brucella spp.
- Add to Blood Agar Base No. 2, Part No. CM0271B; Columbia Blood Agar Base, Part No. CM0331B; or Brucella Medium Base, Part No. CM0169B
- Each vial supplements 500mL of medium
Specifications
Specifications
| Description | Brucella Selective Supplement |
| Form | Freeze-dried |
| Product Type | Selective Supplement |
| Quantity | 10/Pk. |
| Yield | Each vial for 500mL medium |
| Species | Brucella species |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.