Learn More
Thermo Scientific™ Oxoid™ Butzler Campylobacter Selective Supplement
Shop All Thermo Scientific ProductsDescription
Oxoid Campylobacter Selective Supplement, Butzler is used for the isolation of Campylobacter spp. at 37°C.
- Add to Columbia Blood Agar Base, Part No. CM0331B
- Each vial supplements 500mL of medium
Specifications
Specifications
| Description | Campylobacter Selective Supplement, Butzler |
| Form | Freeze-dried |
| Product Type | Selective Supplement |
| Quantity | 10/Pk. |
| Yield | Each vial for 500mL medium |
| Species | Campylobacter |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.