Learn More
Thermo Scientific™ Oxoid™ C-F-C Supplement
Catalog No. OXSR0103E Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
10/Pk.
Catalog No. Quantity
OXSR0103E 10/Pk.
1 options
This item is not returnable. View return policy
Catalog No. OXSR0103E Supplier Thermo Scientific™ Supplier No. SR0103E
Add to Cart
Edge
Add to Cart
This item is not returnable. View return policy
Oxoid C-F-C Supplement is used for the isolation of Pseudomonas species.
Oxoid C-F-C Supplement is used for the isolation of Pseudomonas species.
  • Add to to Pseudomonas Agar Base, Part No. CM0559B
  • Each vial supplements 500mL of medium
TRUSTED_SUSTAINABILITY
Specifications
Description C-F-C Supplement
Form Freeze-dried
Product Type Microbiology Media
Quantity 10/Pk.
Yield Each vial for 500mL medium