Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Oxoid™ C-T Supplement

Catalog No. OXSR0172E Shop All Thermo Scientific Products
Change view
Click to view available options
Yield:
Each vial for 2L medium
Each vial for 500mL medium
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Yield
OXSR0172E Each vial for 500mL medium
OXSR0172H Each vial for 2L medium
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. OXSR0172E Supplier Thermo Scientific™ Supplier No. SR0172E
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Oxoid C-T Supplement is a chromogenic medium for the identification of Escherichia coli O157:H7.

Oxoid C-T Supplement is a chromogenic medium for the identification of Escherichia coli O157:H7.

Add to Sorbitol MacConkey Agar, Part No. CM0813B or to Sorbitol MacConkey Agar with BCIG, Part No. CM0981B.

Ordering Information:

Each vial supplements 2L of medium.

TRUSTED_SUSTAINABILITY

Specifications

Description C-T Supplement
Form Freeze-dried
Product Type Microbiology Media
Quantity 10/Pk.
Yield Each vial for 500mL medium
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.