Learn More
Description
Oxoid Campylobacter Growth Supplement is a liquid supplement for the enhanced growth and aerotolerance of Campylobacter species.
- For the preparation of Blaser-Wang Medium, Bolton Selective Enrichment Broth, Butzler Selective Medium, and Preston Selective Medium
- Each vial supplements 500mL of medium
Specifications
Specifications
| Description | Campylobacter Growth Supplement |
| Form | Freeze-dried |
| Product Type | Microbiology Media |
| Quantity | 10/Pk. |
| Yield | Each vial for 500mL medium |
| Species | Campylobacter |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.