Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Oxoid™ CCDA Selective Supplement

Catalog No. SR0155H Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
10 Vials
10/Pk.
Yield:
Each vial for 500mL medium
For 2L medium
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Yield
SR0155H 10 Vials For 2L medium
OXSR0155E 10/Pk. Each vial for 500mL medium
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. SR0155H Supplier Thermo Scientific™ Supplier No. SR0155H
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Oxoid Brain Heart Infusion Broth is highly nutritious medium for the growth of fastidious organisms, recommended for blood culture work; also see Heart Infusion Broth.

Oxoid CCDA Selective Supplement is used for the isolation of Campylobacter spp., especially C. upsaliensis or Arcobacter.

  • For the isolation of Arcobacter spp., add to Arcobacter Broth Base, Part No. CM0965B
  • For the isolation of Campylobacter spp., especially C. upsaliensis, add to Campylobacter Blood-Free Selective Agar Base (CCDA Preston), Part No. CM0739B
TRUSTED_SUSTAINABILITY

Specifications

Description CCDA Selective Supplement
Form Freeze-dried
Product Type Selective Supplement
Quantity 10 Vials
Yield For 2L medium
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.