Learn More
Thermo Scientific™ Oxoid™ Clostridium Difficile Selective Supplement
Shop All Thermo Scientific ProductsDescription
Oxoid Clostridium Difficile Selective Supplement is added to Clostridium Difficile Agar Base, Part No. CM0601B.
Specifications
Specifications
| Description | Clostridium Difficile Selective Supplement |
| Form | Freeze-dried |
| Product Type | Selective Supplement |
| Quantity | 10/Pk. |
| Yield | Each vial for 500mL medium |