Learn More
Description
Oxoid CO2 Gen Compact Sachet is for use in Compact Plastic Pouch.
- No water or catalyst required
- No hydrogen gas produced
- For use with Compact Plastic Pouch, Part No. AG0020C and AnaeroGen Compact Sealing Clip, Part No. AN0005C
Specifications
Specifications
| Type | CO2 Generating Compact Sachet |
| Format | Compact Gas Generation Sachet |
| Product Line | Thermo Scientific Oxoid |
| Quantity | 20/Pk. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.