Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Oxoid™ Cycloheximide Solution (0.1%)

Catalog No. R663222 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R663222 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. R663222 Supplier Thermo Scientific™ Supplier No. SR0222C
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Oxoid Cycloheximide Solution (0.1%) (100mg/vial) is used for the suppression of fungi.

Oxoid Cycloheximide Solution (0.1%) (100mg/vial) is used for the suppression of fungi.

Add to Bacillus Cereus Selective Agar Base, Part No. CM0617B; Raka-Ray Agar Base, Part No. CM0777B; and Universal Beer Agar, Part No. CM0651B
TRUSTED_SUSTAINABILITY

Specifications

Description Cycloheximide Solution (0.1%)
Form Liquid
Product Type Solution
Quantity 100 mL
Yield Yield will be dependent upon formulation; follow the guidelines for your specific product.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.