Learn More
Thermo Scientific™ Oxoid™ Diagnostic Discs
Catalog No. OXDD0002T Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
50 Discs
Catalog No. Quantity
OXDD0002T 50 Discs
1 options
This item is not returnable. View return policy
Catalog No. OXDD0002T Supplier Thermo Scientific™ Supplier No. DD0002T
This item is not returnable. View return policy
For identification and differentiation of microorganisms
  • Differentiation of Lancefield Group A Streptococci from other beta-haemolytic Streptococci
  • Size: 50 discs
Order Info
Supplied in cartridges of 50 discs each
TRUSTED_SUSTAINABILITY
Specifications
Antimicrobial Agents Bacitracin
Quantity 50 Discs