Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Oxoid™ Dip-Slides™ Industrial

Catalog No. DS0155A Shop All Thermo Scientific Products
Change view
Click to view available options
Format:
Dip Slide
Media Type:
TTC (Red Spot)
TTC (Red Spot)/Malt Ext Agar
Manufacturing Origin:
United Kingdom
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Format Media Type Manufacturing Origin
DS0155A Dip Slide TTC (Red Spot)/Malt Ext Agar United Kingdom
DS0147A Dip Slide TTC (Red Spot) United Kingdom
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. DS0155A Supplier Thermo Scientific™ Supplier No. DS0155A
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Make colony counting simple for aerobic bacteria, yeast, mold, coliform bacteria and Enterobacteriaceae with the Thermo Scientific™ Oxoid™ Dip-Slides™ Industrial.

Make colony counting simple for aerobic bacteria, yeast, mold, coliform bacteria and Enterobacteriaceae with the easily disposable Thermo Scientific™ Oxoid™ Dip-Slides™ Industrial. Simple-to-use for the operator and where laboratory facilities are limited, the Dip Slide is coated on both sides with a solid culture medium with the opportunity for different media to be carried on each side, making the process time- and cost-effective.

Originally designed to determine bacterial counts on urine specimens, dip slides have been modified over the years by making the device double-sided, with one side a medium for Gram positive species and the other side a medium for Gram negatives. With these dip slides, the process for colony counting is time- and cost-effective, allowing exposure of the specimens to two different media simultaneously.

  • Slide is made of easily disposable plastic with raised edges to ensure an even thickness of culture medium
  • Molded grid allows for easier colony counting
  • Slide is shaped to ensure drainage of fluid from the slide
  • Cap forms a convenient handle by which it may be held without risk of touching the culture medium
  • Bacteria, fungal and yeast charts are included, make examining growth simple for the operator and where laboratory facilities are limited

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products with a comprehensive line of media and diagnostic products, providing a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 15°C to 25°C
Format Dip Slide
Manufacturing Origin United Kingdom
Media Type TTC (Red Spot)/Malt Ext Agar
Product Type Dip Slide
Quantity 10/Pk.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.