Learn More
Description
The Oxoid DrySpot™ Pneumo Test is a latex agglutination test for the detection of capsular antigen from Streptococcus pneumoniae to provide rapid identification of Strep. pneumoniae isolated from culture plates and blood culture.
Blue latex particles sensitised with specific rabbit antibody are dried onto the test card. These react with recognised serological types of pneumococci to form agglutination and provide the test reaction area.
Blue latex particles sensitized with non-reactive rabbit globulins are dried onto the test card to provide the control reaction area.
Streptococcus pneumoniae is a primary cause of bacterial pneumonia, meningitis and otitis media. The anti-phagocytic properties of the polysaccharide capsule are the key to the organism’s virulence. The organism may harmlessly inhabit the upper respiratory tract but may also gain access to the lungs by aspiration where it may establish an acute pneumonia. In addition, this organism also accesses the blood stream and the meninges to cause acute, purulent life-threatening infections.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
Specifications
Specifications
| Content And Storage | 2°C to 25°C |
| Control Sets | Positive |
| Description | DrySpot Pneumo Latex Test |
| Type | Streptococcus Test Kit |
| Includes | 20 reaction cards (3 tests/card), positive control, negative control, phosphate buffered saline, mixing paddles, pouch clips |
| Material | Latex |
| Quantity | 60 Tests/Kit |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.