Learn More
Thermo Scientific™ Oxoid™ Fraser Supplement
Catalog No. OXSR0156E Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
10/Pk.
Catalog No. Quantity
OXSR0156E 10/Pk.
1 options
This item is not returnable. View return policy
Catalog No. OXSR0156E Supplier Thermo Scientific™ Supplier No. SR0156E
Add to Cart
Add to Cart
This item is not returnable. View return policy
Oxoid Fraser Supplement is used for the isolation of Listeria species.
Oxoid Fraser Supplement is used for the isolation of Listeria species.
  • Add to Fraser Broth Base, Part No. CM0895B
  • Each vial supplements 500mL of medium
TRUSTED_SUSTAINABILITY
Specifications
Description Fraser Supplement
Form Freeze-dried
Product Type Microbiology Media
Quantity 10/Pk.
Yield Each vial for 500mL medium