Learn More
Thermo Scientific™ Oxoid™ Gelatin Bacteriological
Catalog No. OXLP0008B Shop All Thermo Scientific Products
Change view
Click to view available options
Description:
Dehydrated; Gelatin bacteriological
Catalog No. Description
OXLP0008B Dehydrated; Gelatin bacteriological
1 options
This item is not returnable. View return policy
Catalog No. OXLP0008B Supplier Thermo Scientific™ Supplier No. LP0008B
Add to Cart
Add to Cart
This item is not returnable. View return policy
Oxoid Gelatin Bacteriological is a special bacteriological grade readily soluble in water to give a clear solution free from sulphite and preservatives.
  • Firm gel obtained from 15% solution
TRUSTED_SUSTAINABILITY
Specifications
Description Dehydrated; Gelatin bacteriological
Form Powder
Product Type Gelatin
Quantity 500 g