Learn More
Description
Oxoid Half Fraser Supplement is used for the enrichment of Listeria species.
- Add to Fraser Broth Base, Part No. CM0895B
Specifications
Specifications
| Description | Half Fraser Supplement |
| Form | Freeze-dried |
| Product Type | Microbiology Media |
| Quantity | For 2.25L of base |
| Species | Listeria |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.