Learn More
Thermo Scientific™ Oxoid™ Helicobacter Pylori Selective Supplement (Dent)
Shop All Thermo Scientific ProductsDescription
Oxoid Helicobacter Pylori Selective Supplement (Dent) is for the isolaton of Helicobacter pylori from clinical specimens.
- Used with Columbia Blood Agar Base, Part No. CM0331B
- Each vial supplements 500mL of medium
Specifications
Specifications
| Description | Helicobacter Pylori Selective Supplement (Dent) |
| Form | Freeze-dried |
| Product Type | Selective Supplement |
| Quantity | 10/Pk. |
| Yield | Each vial for 500mL medium |
| Species | Helicobacter pylori |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.