Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Oxoid™ Horse Serum

Catalog No. oxsr0035c
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
OXSR0035C 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. OXSR0035C Supplier Thermo Scientific™ Supplier No. SR0035C
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Thermo Scientific™ Oxoid Horse Serum is an extract of horse blood for addition to culture media.

Thermo Scientific™ Oxoid Horse Serum is an extract of horse blood for addition to culture media.

TRUSTED_SUSTAINABILITY

Specifications

Species Horse
Form Liquid
Product Type Horse Whole Blood, Supplement
Quantity 100 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.