Learn More
Thermo Scientific™ Oxoid™ IMAGEN™ Chlamydia Positive Control Slide
Catalog No. RS6109302 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
1 Slide/Pk.
Catalog No. Quantity
RS6109302 1 Slide/Pk.
1 options
This item is not returnable. View return policy
Catalog No. RS6109302 Supplier Thermo Scientific™ Supplier No. S6109302
Add to Cart
Add to Cart
This item is not returnable. View return policy
Oxoid IMAGEN Chlamydia Positive Control Slide contains acetone-fixed human epithelial cells infected with Chlamydia.
The IMAGEN™ Chlamydia Control Slides are for use as a control for the IMAGEN™ Chlamydia test (Code No. K610111-2).
Sealed in foil pouch filled with nitrogen.
TRUSTED_SUSTAINABILITY
Specifications
Description IMAGEN™ Chlamydia Positive Control Slide
Detectable Analytes Chlamydia species
Type Chlamydia Control Slide
Quantity 1 Slide/Pk.