Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ IMAGEN™ Parainfluenza Virus Group Kit using Direct Immunofluorescence Assay

Catalog No. K6103112 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
50 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
K6103112 50 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. K6103112 Supplier Thermo Scientific™ Supplier No. K6103112
 Restricted Item
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Rapidly detect Parainfluenza virus types 1, 2 and 3 with this rapid and easy-to-read kit.

Rapidly detect Parainfluenza virus types 1, 2 and 3 antigens directly in nasopharyngeal aspirates or in cell culture preparations with Thermo Scientific™ IMAGEN™ Parainfluenza Group Kit. This direct immunofluorescence assay utilizes type-specific monoclonal antibodies to detect and identify Parainfluenza virus types 1, 2 and 3.

Parainfluenza viruses are members of the genus Paramyxovirus classified within the family Paramyxoviridae.1 There are 6 species of Paramyxovirus known to infect hummans which include Parainfluenza viruses 1, 2, 3, and 4 (subtypes 4a and 4b), Mumps virus, and Newcastle Disease virus. Of the 4 Parainfluenza viruses, types 1, 2, and 3 are now recognized as a major cause of acute respiratory disease in infants and children.2,3

Use IMAGEN Parainfluenza Kit as a rapid, sensitive, and specific method for detection of Parainfluenza virus types 1, 2, and 3 antigens directly in nasopharyngeal aspirates or in cell culture preparations.

  • Rapid results—within a little over 30 minutes of receiving test sample or culture
  • Simple—ready-to-use reagents plus all IMAGEN assays follow the same basic methodology
  • Excellent sensitivity and specificity—high quality, well-characterized antibodies used in the assay provide excellent sensitivity and specificity
  • Easy-to-read results
  • Long shelf life—24 months

The IMAGEN Parainfluenza Kit contains fluorescein isothiocyanate (FITC) conjugated monoclonal antibodies which bind specifically to either Parainfluenza virus types 1, 2, or 3. Specimens are incubated with the FITC conjugated reagents for 15 minutes. Excess reagent is then removed by washing with phosphate-buffered saline (PBS). The stained areas are mounted and viewed using epifluorescence illumination. If Parainfluenza virus types 1, 2, or 3 are present then characteristic apple-green intracellular fluorescence is seen within infected cells, which contrasts with the red background staining of uninfected cells.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 2°C to 8°C
Description IMAGEN™ Parainfluenza Group
Detectable Analytes Influenza
Sample Type Nasopharyngeal
Type Reagent
Quantity 50 Tests
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.