Description
Oxoid LCAT Selective Supplement is used for the preparation of New York City Medium for the isolation of Neisseria spp.
- Add to GC Agar Base, Part No. CM0367B
- Each vial supplements 500mL of medium
Specifications
Specifications
| Description | Lcat Select Suppl |
| Format | Freeze Dried |
| Product Type | Selective Supplement |
| Quantity | 10/Pk. |
| Yield | Each vial for 500mL medium |