Learn More
Thermo Scientific™ Oxoid™ Malt Extract
Catalog No. oxlp0039b
Change view
Click to view available options
Quantity:
500 g
Catalog No. Quantity
OXLP0039B 500 g
1 options
This item is not returnable. View return policy
Catalog No. OXLP0039B Supplier Thermo Scientific™ Supplier No. LP0039B
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy
Oxoid Malt Extract is recommended for use in media for yeast and mold growth.
An extract of sprouted grain with conserved nitrogen and carbohydrate constituents
TRUSTED_SUSTAINABILITY
Specifications
Description Dehydrated; Malt extract
Form Powder
Product Type Malt Extract
Quantity 500 g