Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Oxoid™ Meropenem/Vaborbactam MEV30 Antimicrobial Susceptibility Discs

Catalog No. STCT1949B
Change view
Click to view available options
Concentration:
20/10 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Concentration
STCT1949B 20/10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. STCT1949B Supplier Thermo Scientific™ Supplier No. CT1949B
Add to Cart
Add to Cart
This item is not returnable. View return policy

Test the susceptibility of carbapenem-resistant Enterobacteriaceae to meropenem-vaborbactam with easy-to-use AST disks that meet CLSI breakpoints

Reliably and easily determine the antibiotic susceptibility of carbapenem-resistant - Enterobacteriaceae (CRE) using manual disk diffusion Antimicrobial Susceptibility Testing (AST) methods with Thermo Scientific™ Oxoid™ Meropenem/Vaborbactam MEV30 Antimicrobial Susceptibility Discs.Meropenem/Vaborbactam MEV30 Discs offer a direct resistance comparison of meropenem to meropenem/vaborbactam to help physicians understand the clinical benefit of vaborbactam as a treatment option for serious Gram-negative infections, such as complicated urinary tract infections (cUTIs).

Perform susceptibility testing of carbapenem-resistant Enterobacteriaceae (CRE) using Oxoid Meropenem/Vaborbactam MEV30 Antimicrobial Susceptibility Discs which contain the carbapenem, meropenem, and the novel beta-lactamase inhibitor, vaborbactam. The easy-to-use disk format may assist in the decision-making process of adding meropenem/ vaborbactam into your formulary.
  • Meets CLSI recommended breakpoints
  • Easy-to-use: shatterproof spring-loaded cartridges and easy-to-read labeling.
  • Long-term stability ensured: each cartridge is individually sealed with a desiccant to maintain a moisture level of <2%.
  • Accurate and reproducible: manufactured under stringent quality standards including BS EN ISO 9001:2008, ISO 13485:2003 and FDA 21 CFR part 820, with comprehensive quality control testing.
  • Qualitative: categorizes resistant, intermediate, and sensitive organisms
  • Trusted supplier: over 35 years of experience in disk development and manufacture

Meropenem/vaborbactam has been shown to be active both clinically and in vitro against:

  • Escherichia coli
  • Klebsiella pneumoniae
  • Enterobacter cloacae species complex

Disk diffusion not only categorizes resistant, intermediate, and sensitive organisms through qualitative results but also provides a visual indication of the following:

  • Inoculum level
  • Presence of contamination
  • Resistant mutants
  • Beta-lactamase activity
  • Antagonism and synergy between antibiotics

Thermo Fisher Scientific is a leader in providing AST solutions that help in the fight against antimicrobial resistance, offering high performance automated and manual test options and one of the largest, most up-to-date selections of antimicrobials for susceptibility testing.

TRUSTED_SUSTAINABILITY

Specifications

Antimicrobial Agents Meropenem/Vaborbactam
Concentration 20/10 μg
Description OxoidTM Meropenem/Vaborbactam Antimicrobial Susceptibility Discs
Antibiotic Code MEV
Product Type Antimicrobial Susceptibility Discs
Target Organism Class Gram-negative Microorganisms
Test Type Antimicrobial Susceptibility
No. per Pack 5 Cartridges/Pk.
Packaging Type 50 Discs/Cartridge
Quantity 50 Discs/Cartridge
Species Carbapenem-Resistant Enterobacteriaceae (CRE)
Show More Show Less

Not all products are available for sale in all territories. Please inquire. Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.