Learn More
Thermo Scientific™ Oxoid™ Microbact™ GNB Reagents and Accessories
Shop All Thermo Scientific ProductsDescription
Simplify the detection of Gram-negative bacteria with convenient Thermo Scientific™ Oxoid™ Microbact™ GNB Reagents and Accessories.
- Fast: More results available overnight
- Simple: Simple test strip or microplate format
- Easy to Read and Interpret: Results are clearly visible as distinct color reactions that can be interpreted using the Microbact Identification Package (MB1244A)
- Comprehensive: Identifies >100 Gram-negative bacteria, more unusual Enterobacteriaceae and oxidase-positive, Gram-negative bacteria
- Flexible: Use single strips or combine systems for more complex identifications. Ideal for use in clinical or food microbiology laboratories
- Reliable: A comprehensive range of biochemical tests based on published reference methodologies1-4
Additional items required: Mineral Oil (MB1093A), Oxidase Strips (MB0266A), Microbact Identification Package (Windows™) (MB1244A), Indole Reagent (MB0209A), VPI Reagent (MB0181A), VPII Reagent (MB0184A), TDA Reagent (MB0180A), NIT A Reagent (MB0186A), NIT B Reagent (MB0187A). The 6 reagents above are also available as Reagent Set D (MB1082A).
Not all products are available for sale in all territories. Please inquire.
Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products with a comprehensive line of media and diagnostic products, providing a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
Specifications
Specifications
| Description | Microbact Mineral Oil |
| Format | Bottle |
| Packaging Type | 1 Box |
| Quantity | 1 Bottle |